Latanoprost

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Latanoprost, (Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R)-3-hydroxy-5-phenylpentyl)cyclopentyl)hept-5-enoate

46666-0250 46666-1000
TS46666-0250EA 156 USD
TS46666-0250 TS46666-1000
Latanoprost
Latanoprost
Formula: C₂₆H₄₀O₅
MW: 432.60 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00216074
CAS Number: 130209-82-4
UN: 2810
ADR: 6.1,III
Merck Index: 13.5391

Order Now

Specification Test Results

Appearance Clear colorless to light yellow liquid
Infrared spectrum Conforms
HPLC ≥ 98.5%
Water ≤ 0.5%
Residue after ignition ≤ 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR