Genipin

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate, (1R,4aS,7aS)-Methyl 1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate

46664-0010 46664-2500
TS46664-0010EA 340 USD
TS46664-0010 TS46664-2500
Genipin
Genipin
Formula: C₁₁H₁₄O₅
MW: 226.23 g/mol
Melting Pt: 121°C
Storage Temperature: Refrigerator
MDL Number: MFCD00888600
CAS Number: 6902-77-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to almost white powder
Infrared spectrum Conforms
Melting point 118.0°C to 123.0°C
HPLC ≥ 98.0%
Water ≤ 1.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR