Flutamide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(4-Nitro-3-(trifluoromethyl)phenyl)isobutyramide, 4'-Nitro-3'-(trifluoromethyl)isobutyranilide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide

46645-0050 466450250
TS46645-0050EA 100 USD
TS46645-0050 TS466450250
Flutamide
Flutamide
Formula: C₁₁H₁₁F₃N₂O₃
MW: 276.22 g/mol
Melting Pt: 112°C
Storage Temperature: Ambient
MDL Number: MFCD00072009
CAS Number: 13311-84-7
EINECS: 236-341-9
Merck Index: 13.4234

Order Now

Specification Test Results

Appearance White to yellow powder
Infrared spectrum Conforms
Melting point 110.0°C to 114.0°C
HPLC ≥ 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR