N-(Phosphonomethyl)glycine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glyphosate

46618-0010 46618-0050
TS46618-0010EA 95 USD
TS46618-0010 TS46618-0050
N-(Phosphonomethyl)glycine
N-(Phosphonomethyl)glycine
Formula: C₃H₈NO₅P
MW: 169.07 g/mol
Melting Pt: 230°C
Storage Temperature: Ambient
MDL Number: MFCD00055350
CAS Number: 1071-83-6
EINECS: 213-997-4
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to light yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥ 94%
Water ≤ 1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR