Gemifloxacin mesylate

Supplier: Thermo Scientific Chemicals

46597-2500 465970010
TS46597-2500EA 310 USD
TS46597-2500 TS465970010
Gemifloxacin mesylate
Gemifloxacin mesylate
Formula: C₁₈H₂₀FN₅O₄·CH₄O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD08277637
CAS Number: 210353-53-0

Order Now

Specification Test Results

Appearance White to yellow to brown powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Water ≤ 10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR