Bedaquiline fumarate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1R,2S)-1-(6-Bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol fumarate

46591-0010 46591-2500
TS46591-0010EA 634 USD
TS46591-0010 TS46591-2500
Bedaquiline fumarate
Bedaquiline fumarate
Formula: C₃₆H₃₅BrN₂O₆
MW: 671.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28167761
CAS Number: 845533-86-0

Order Now

Specification Test Results

Appearance White to almost white powder
Infrared spectrum Conforms
Water ≤ 1%
HPLC ≥ 98.5%
Fumaric acid 17.0 to 17.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR