Proflavine hydrochloride (3,6-diaminoacridine hydrochloride)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acridine-3,6-diamine hydrochloride

46586-0050 465860250
TS46586-0050EA 50 USD
TS46586-0050 TS465860250
Proflavine hydrochloride (3,6-diaminoacridine hydrochloride)
Proflavine hydrochloride (3,6-diaminoacridine hydrochloride)
Formula: C₁₃H₁₁N₃·HCl
MW: 245.71 g/mol
Melting Pt: 270°C
Storage Temperature: Ambient
MDL Number: MFCD00066867
CAS Number: 952-23-8
EINECS: 213-469-9

Order Now

Specification Test Results

Appearance Dark orange to red-brown powder
Infrared spectrum Conforms
Titration Argentometry 13.6 to 15.0% (Cl)
HPLC ≥ 94%
Loss on drying ≤ 4% (105°C, 1hr)
Solubility (1mg/ml in water) Clear solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR