β-Methasone sodium phosphate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: β-Methasone 21-phosphate disodium salt, Betamethasone sodium phosphate

46574-0010 46574-0050
TS46574-0010EA 118 USD
TS46574-0010 TS46574-0050
β-Methasone sodium phosphate
β-Methasone sodium phosphate
Formula: C₂₂H₂₈FO₈P·₂Na
MW: 516.41 g/mol
Melting Pt: 206°C…211°C
Storage Temperature: Refrigerator
MDL Number: MFCD00200361
CAS Number: 151-73-5
EINECS: 205-797-0

Order Now

Specification Test Results

Appearance White to almost white powder
Infrared spectrum Conforms
Water ≤ 10%
HPLC ≥ 96.0%
Specific optical rotation +98° to +105° (20°C, 589nm) (1% in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR