Sorafenib tosylate (BAY 43-9006 tosylate salt)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, 4-methylbenzenesulphonate, Sorafenib Tosylate, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-methylbenzenesulphonate, 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)phenoxy)-N-methylpicolinamide 4-methylbenzenesulphonate

46571-0010 46571-2500
TS46571-0010EA 296 USD
TS46571-0010 TS46571-2500
Sorafenib tosylate (BAY 43-9006 tosylate salt)
Sorafenib tosylate (BAY 43-9006 tosylate salt)
Formula: C₂₁H₁₆ClF₃N₄O₃·C₇H₈O₃S
MW: 637.03 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08235032
CAS Number: 475207-59-1

Order Now

Specification Test Results

Appearance Almost white to light brown powder
Infrared spectrum Conforms
HPLC ≥ 97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR