Pomalidomide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-N-(2,6-dioxo-3-piperidyl)phthalimide, 4-Amino-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

46570-0010 46570-2500
TS46570-0010EA 408 USD
TS46570-0010 TS46570-2500
Pomalidomide
Pomalidomide
Formula: C₁₃H₁₁N₃O₄
Melting Pt: 315°C…320°C
Storage Temperature: Refrigerator
MDL Number: MFCD12756407
CAS Number: 19171-19-8

Order Now

Specification Test Results

Appearance Yellow to green powder
Infrared spectrum Conforms
HPLC ≥ 98.5%
Loss on drying ≤ 1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR