Bosutinib

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: SKI-606

46565-2500 465650010
TS46565-2500EA 175 USD
TS46565-2500 TS465650010
Bosutinib
Bosutinib
Formula: C₂₆H₂₉Cl₂N₅O₃
MW: 530.5 g/mol
Melting Pt: 131°C…134°C
Storage Temperature: Ambient
MDL Number: MFCD07367846
CAS Number: 380843-75-4

Order Now

Specification Test Results

Appearance Almost white to yellow crystalline powder
Infrared spectrum Conforms
Loss on drying ≤ 0.5%
HPLC ≥ 97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR