Afatinib 96%

Supplier: Thermo Scientific Chemicals
Warning

46564-2500 465640010
TS46564-2500EA 120 USD
TS46564-2500 TS465640010
Afatinib 96%
Afatinib
Formula: C₂₄H₂₅ClFN₅O₃
Melting Pt: 102°C
Storage Temperature: Ambient
MDL Number: MFCD12407405
CAS Number: 850140-72-6

Order Now

Specification Test Results

Appearance White to beige powder
Infrared spectrum Conforms
Water ≤ 5.0%
HPLC ≥ 95.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR