Fipronil

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-3-cyano-4-(trifluoromethyl)sulphinylpyrazole, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)sulphinylpyrazole-3-carbonitrile

46557-0050 46557-0250 46557-1000
TS46557-0050EA 64 USD
TS46557-0050 TS46557-0250 TS46557-1000
Fipronil
Fipronil
Formula: C₁₂H₄Cl₂F₆N₄OS
MW: 437.14 g/mol
Melting Pt: 200°C…201°C
Storage Temperature: Refrigerator
MDL Number: MFCD00867812
CAS Number: 120068-37-3
UN: 2811
ADR: 6.1,III
Merck Index: 13.4115

Order Now

Specification Test Results

Appearance White to yellow to beige powder or crystals
Infrared spectrum Conforms
Melting point 194°C to 205°C
HPLC ≥ 97.5%
Loss on drying ≤ 2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR