Valsartan

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid, (S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid

46556-0010 46556-0050
TS46556-0010EA 46 USD
TS46556-0010 TS46556-0050
Valsartan
Valsartan
Formula: C₂₄H₂₉N₅O₃
MW: 435.52 g/mol
Melting Pt: 117°C
Storage Temperature: Refrigerator
MDL Number: MFCD00865840
CAS Number: 137862-53-4
Merck Index: 13.9982

Order Now

Specification Test Results

Appearance White to almost white to light beige powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Specific optical rotation -55° to -70° (20°C, 589nm) (1% in CH3OH)
Water ≤ 2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR