5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5,5'-Diallylbiphenyl-2,2'-diol, 5,5'-Diallyl-[1,1'-biphenyl]-2,2'-diol, Magnolol
46551-2500
465510010
TS46551-2500EA
99
USD
TS46551-2500
TS465510010
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
Formula:
C₁₈H₁₈O₂ Melting Pt: 101°C…102°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016658 CAS Number: 528-43-8 |
Specification Test Results
Appearance | White to yellow to beige powder |
Infrared spectrum | Conforms |
Melting point | 99°C to 103°C |
Water | ≤ 1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...