Parecoxib Sodium

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dynastat, SC-69124A, Sodium ((4-(5-methyl-3-phenylisoxazol-4-yl)phenyl)sulphonyl)(propionyl)amide

46538-0010 465380050
TS46538-0010EA 318 USD
TS46538-0010 TS465380050
Parecoxib Sodium
Parecoxib sodium
Formula: C₁₉H₁₇N₂NaO₄S
MW: 392.4 g/mol
Melting Pt: 273°C…275°C
Storage Temperature: Refrigerator
MDL Number: MFCD04972608
CAS Number: 198470-85-8

Order Now

Specification Test Results

Appearance White to beige powder
Infrared spectrum Conforms
HPLC ≥ 98.5%
Water ≤ 3%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR