Aprepitant

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(((2R,3S)-2-((R)-1-(3,5-Bis(trifluoromethyl)phenyl)ethoxy)-3-(4-fluorophenyl)morpholino)methyl)-1H-1,2,4-triazol-3(2H)-one

46531-0010 46531-2500
TS46531-0010EA 545 USD
TS46531-0010 TS46531-2500
Aprepitant
Aprepitant
Formula: C₂₃H₂₁F₇N₄O₃
Melting Pt: 255°C
Storage Temperature: Ambient
MDL Number: MFCD08277635
CAS Number: 170729-80-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to almost white powder
Infrared spectrum Conforms
Melting point 250°C to 255°C
Water ≤ 0.5%
HPLC ≥ 97.5%
Specific optical rotation +66° to +71° (20°C, 589nm) (1% in M%eOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR