Azelnidipine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester, 3-(1-Benzhydrylazetidin-3-yl) 5-isopropyl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

46506-0010 465060050
TS46506-0010EA 70 USD
TS46506-0010 TS465060050
Azelnidipine
Azelnidipine
Formula: C₃₃H₃₄N₄O₆
MW: 582.66 g/mol
Melting Pt: 193.3°C…195°C
Storage Temperature: Ambient
MDL Number: MFCD00865803
CAS Number: 123524-52-7

Order Now

Specification Test Results

Appearance Light yellow to yellow powder
Infrared spectrum Conforms
Melting point 120°C to 126°C
HPLC ≥ 97.5%
Loss on drying ≤ 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR