Hydrocortisone 17-Butyrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 11beta,17,21-Trihydroxypregn-4-ene-3,20-dione 17-Butyrate, Cortisol 17-Butyrate

46505-0010 465050050 465052500
TS46505-0010EA 104 USD
TS46505-0010 TS465050050 TS465052500
Hydrocortisone 17-Butyrate
Hydrocortisone 17-Butyrate
Formula: C₂₅H₃₆O₆
MW: 432.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083364
CAS Number: 13609-67-1
EINECS: 237-093-4
Merck Index: 13.4808

Order Now

Specification Test Results

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
Loss on drying ≤ 1%
HPLC ≥ 96.0%
Specific optical rotation +47° to +54° (20°C, 589nm) (0.1% in CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR