Rocuronium bromide 97%

Supplier: Thermo Scientific Chemicals
Warning

46502-0010 46502-2500
TS46502-0010EA 335 USD
TS46502-0010 TS46502-2500
Rocuronium bromide 97%
Rocuronium bromide
Formula: C₃₂H₅₃BrN₂O₄
MW: 609.69 g/mol
Melting Pt: 162°C…164°C
Storage Temperature: Freezer
MDL Number: MFCD00867768
CAS Number: 119302-91-9

Order Now

Specification Test Results

Appearance White to light yellow to light brown crystalline powder
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0% (dry substance)
Loss on drying ≤ 6%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR