β-Methasone valerate

Supplier: Thermo Scientific Chemicals
Danger

46493-0010 46493-0050
TS46493-0010EA 126 USD
TS46493-0010 TS46493-0050
β-Methasone valerate
beta-Methasone valerate
Formula: C₂₇H₃₇FO₆
MW: 476.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00867446
CAS Number: 2152-44-5
EINECS: 218-439-3

Order Now

Specification Test Results

Appearance White to almost white powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Loss on drying ≤ 0.5% (1g,105°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR