Bendazac

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [(1-Benzyl-3-indazolyl)oxy]acetic Acid, Bendazolic acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole

46492-0010 464920050
TS46492-0010EA 65 USD
TS46492-0010 TS464920050
Bendazac
Bendazac
Formula: C₁₆H₁₄N₂O₃
MW: 282.30 g/mol
Melting Pt: 160°C
Storage Temperature: Ambient
MDL Number: MFCD00866158
CAS Number: 20187-55-7
EINECS: 243-569-2
Merck Index: 13.1032

Order Now

Specification Test Results

Appearance White powder
Infrared spectrum Conforms
Melting point 160°C to 164°C
HPLC ≥ 97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR