Chromium etchant 1020AC semiconductor grade

Supplier: TRANSENE PC
Warning

0021001-GAL 0021001-QT
76541-012CS 482.59 USD
76541-012 76541-018
Chromium etchant 1020AC semiconductor grade
Chromium etchant

Chromium Etchant 1020AC is a slower etchant for chrome thin films or photomasks. Slower etch rate provides better control, enhanced fine-line definition, and compatibility with a broader range of materials and photoresists.

Order Now

Specification Test Results

Appearance Clear orange
Etch Capacity (rate declines at ~70%) 65 g/gallon
Etch Rate at 40°C 32/sec Å/sec
Filtration 0,2 um
Incompatible Materials Al, Ni, Cu, NiCr
pH Acidic
Recommended Operating Temperatures 20-80°C (30-40°C most common) °C
Shelf Life 1 year
Storage Conditions Ambient


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR