Nitric acid 69.5-70%, Low-Metal (LM) grade semiconductor grade

Supplier: TRANSENE PC
Danger

Synonyms: Nitric acid solution

060-00060-QT 060-00060-7LB
76540-970EA 24 USD
76540-970 76541-032
Nitric acid 69.5-70%, Low-Metal (LM) grade semiconductor grade
Nitric acid

Transene Nitric Acid LM is a "low metals" nitric acid solution suitable for demanding semiconductor and electronics processing.

Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 122 °C
Melting Pt: –42 °C
Density: 1.41 g/cc
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8, 5.1,II

Order Now

Specification Test Results

Assay (HNO3) 69-70 %
Color 10 APHA max.
Chloride (Cl) 80 ppb max.
Residue After Ignition 2 ppm max.
Aluminum (Al) 20 ppb max.
Arsenic and Antimony (as As) 5 ppb max.
Barium (Ba) 10 ppb max.
Beryllium (Be) 10 ppb max.
Bismuth (Bi) 10 ppb max.
Boron (B) 10 ppb max.
Cadmium (Cd) 10 ppb max.
Calcium (Ca) 50 ppb max.
Chromium (Cr) 10 ppb max.
Cobalt (Co) 10 ppb max.
Copper (Cu) 10 ppb max.
Gallium (Ga) 10 ppb max.
Germanium (Ge) 10 ppb max.
Gold (Au) 10 ppb max.
Iron (Fe) 50 ppb max.
Lead (Pb) 10 ppb max.
Lithium (Li) 10 ppb max.
Magnesium (Mg) 10 ppb max.
Manganese (Mn) 10 ppb max.
Molybdenum (Mo) 10 ppb max.
Nickel (Ni) 10 ppb max.
Phosphate (PO4) 200 ppb max.
Platinum (Pt) 10 ppb max.
Potassium (K) 20 ppb max.
Silver (Ag) 10 ppb max.
Sodium (Na) 50 ppb max.
Strontium (Sr) 10 ppb max.
Sulfate (SO4) 500 ppb max.
Thallium (Tl) 10 ppb max.
Tin (Sn) 10 ppb max.
Titanium (Ti) 10 ppb max.
Vanadium (V) 10 ppb max.
Zinc (Zn) 10 ppb max.
Zirconium (Zr) 10 ppb max.
Particle Count, 0.5 mm 50 ct/ml max.
Particle Count, 1.0 mm 10 ct/ml max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR