2-Propanol ≥99.80%, Low-Metal (LM) grade

Supplier: TRANSENE PC
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

001100LM-GAL 001100LM-QT
76540-938CS 133 USD
76540-938 76541-042
2-Propanol ≥99.80%, Low-Metal (LM) grade
2-Propanol

Transene Isopropyl Alcohol LM (synonyms include IPA, 2-propanol, and isopropanol) is a "low metals" alcohol suitable for demanding semiconductor and electronics processing. Effective for cleaning, degreasing, and other applications. Low water content.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C
Melting Pt: –89 °C
Density: 0.8 g/cc
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

Assay (C3H8O) Min 99.50 %
Assay (H2O) 500 ppm max.
Color 6 APHA max.
Appearance Pass/Fail
Acidity 0.200 ueq/g max.
Alkalinity 0.100 ueq/g max.
Residue after Evaporation 5000 ppb max.
Solubility (in H2O) Pass/Fail
Aluminum (Al) 50 ppb max.
Arsenic and Antimony (as As) 10 ppb max.
Barium (Ba) 20 ppb max.
Beryllium (Be) 10 ppb max.
Bismuth (Bi) 20 ppb max.
Boron (B) 10 ppb max.
Cadmium (Cd) 10 ppb max.
Calcium (Ca) 10 ppb max.
Chloride (Cl) 200 ppb max.
Chromium (Cr) 10 ppb max.
Cobalt (Co) 10 ppb max.
Copper (Cu) 10 ppb max.
Gallium (Ga) 10 ppb max.
Germanium (Ge) 10 ppb max.
Gold (Au) 20 ppb max.
Iron (Fe) 50 ppb max.
Lead (Pb) 10 ppb max.
Lithium (Li) 10 ppb max.
Magnesium (Mg) 20 ppb max.
Manganese (Mn) 10 ppb max.
Molybdenum (Mo) 10 ppb max.
Nickel (Ni) 10 ppb max.
Niobium (Nb) 10 ppb max.
Phosphate (PO4) 500 ppb max.
Platinum 50 ppb max.
Potassium (K) 20 ppb max.
Silicon (Si) 100 ppb max.
Silver (Ag) 10 ppb max.
Sodium (Na) 100 ppb max.
Strontium (Sr) 10 ppb max.
Tantalum (Ta) 10 ppb max.
Thallium (Tl) 10 ppb max.
Tin (Sn) 20 ppb max.
Titanium (Ti) 20 ppb max.
Vanadium (V) 10 ppb max.
Zinc (Zn) 20 ppb max.
Zirconium (Zr) 20 ppb max.
Particle Count, 0.5 um 17 ct/ml max.
Particle Count, 1.0 um 5 ct/ml max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR