Hydrochloric acid 36.5-38%, Low-Metal (LM) grade semiconductor grade

Supplier: TRANSENE PC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

HCL0000-GAL HCL0000-QT
76541-028CS 146 USD
76541-028 76541-062
Hydrochloric acid 36.5-38%, Low-Metal (LM) grade semiconductor grade
Hydrochloric acid

Transene Hydrochloric Acid LM is a "low metals" HCl solution suitable for demanding semiconductor and electronics processing.

Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 109 °C
Melting Pt: –74 °C
Density: 1.16 g/cc
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay (HCl) 36.5-38.0 %
Ammonium (NH4) 500 ppb max.
Color 7 APHA max.
Phosphate (PO4) 50 ppb max.
Free Chloride (Cl) 500 ppb max.
Heavy Metals 100 ppb max.
Bromide (Br) 50000 ppb max.
Extractable Organics 500 ppb max.
Residue After Ignition 3000 ppb max.
Aluminum (Al) 10 ppb max.
Arsenic and Antimony (as As) 5 ppb max.
Barium (Ba) 10 ppb max.
Beryllium (Be) 10 ppb max.
Bismuth (Bi) 10 ppb max.
Boron (B) 10 ppb max.
Calcium (Ca) 5 ppb max.
Chromium (Cr) 10 ppb max.
Cobalt (Co) 10 ppb max.
Copper (Cu) 10 ppb max.
Gallium (Ga) 10 ppb max.
Germanium (Ge) 10 ppb max.
Gold (Au) 10 ppb max.
Indium (In) 10 ppb max.
Iron (Fe) 10 ppb max.
Lead (Pb) 10 ppb max.
Lithium (Li) 10 ppb max.
Magnesium (Mg) 10 ppb max.
Manganese (Mn) 10 ppb max.
Molybdenum (Mo) 10 ppb max.
Nickel (Ni) 10 ppb max.
Platinum (Pt) 10 ppb max.
Potassium (K) 10 ppb max.
Silver (Ag) 10 ppb max.
Sodium (Na) 10 ppb max.
Strontium (Sr) 10 ppb max.
Sulfate (SO4) 500 ppb max.
Sulfite (SO3) 800 ppb max.
Titanium (Ti) 10 ppb max.
Zinc (Zn) 10 ppb max.
Zirconium (Zr) 10 ppb max.
Particle Count, 0.5 mm 50 ct/ml max.
Particle Count, 1.0 mm 10 ct/ml max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR