Acetone ≥99.90%, Low-Metal (LM) grade semiconductor grade

Supplier: TRANSENE PC
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

ACETONELM-GAL ACETONELM-QT
76541-024CS 155 USD
76541-024 76541-058
Acetone ≥99.90%, Low-Metal (LM) grade semiconductor grade
Acetone

Transene Acetone LM grade is a "low metals" solvent suitable for electronics and semiconductor cleaning and processing. Low water content.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.5 °C
Melting Pt: –95 °C
Density: 0.8 g/cc
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II

Order Now

Specification Test Results

Assay (Acetone) Min 99.50 %
Isopropyl Alcohol (IPA) 0.05 % max.
Assay (H2O) 0.20 % max.
Assay (C3H8O) 0.05 % max.
Assay (CH3OH) 0.05 % max.
Color 10 APHA max.
Appearance Pass/Fail
Acidity 0.300 ueq/g max.
Alkalinity 0.500 ueq/g max.
Aldehydes 10 ppb (umg/g) max.
Residue after Evaporation 5000 ppb max.
Solubility (in H2O) Pass/Fail
Aluminum (Al) 50 ppb max.
Arsenic and Antimony (as As) 10 ppb max.
Barium (Ba) 10 ppb max.
Beryllium (Be) 10 ppb max.
Bismuth (Bi) 20 ppb max.
Boron (B) 10 ppb max.
Cadmium (Cd) 10 ppb max.
Calcium (Ca) 100 ppb max.
Chloride (Cl) 200 ppb max.
Chromium (Cr) 10 ppb max.
Cobalt (Co) 10 ppb max.
Copper (Cu) 10 ppb max.
Gallium (Ga) 10 ppb max.
Germanium (Ge) 10 ppb max.
Gold (Au) 20 ppb max.
Iron (Fe) 50 ppb max.
Lead (Pb) 10 ppb max.
Lithium (Li) 10 ppb max.
Magnesium (Mg) 20 ppb max.
Manganese (Mn) 10 ppb max.
Molybdenum (Mo) 10 ppb max.
Nickel (Ni) 10 ppb max.
Niobium (Nb) 10 ppb max.
Phosphate (PO4) 100 ppb max.
Platinum (Pt) 50 ppb max.
Potassium (K) 20 ppb max.
Silicon (Si) 100 ppb max.
Silver (Ag) 10 ppb max.
Sodium (Na) 100 ppb max.
Strontium (Sr) 10 ppb max.
Sulfate (SO4) 200 ppb max.
Tantalum (Ta) 10 ppb max.
Thallium (Tl) 10 ppb max.
Tin (Sn) 20 ppb max.
Titanium (Ti) 20 ppb max.
Vanadium (V) 10 ppb max.
Zinc (Zn) 20 ppb max.
Zirconium (Zr) 20 ppb max.
Particle Count, 0.5 um 50 ct/ml max.
Particle Count, 1.0 um 10 ct/ml max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR