Acridine orange base ≥75% (dye basis), Sigma-Aldrich®


M-Clarity™ quality level = MQ200

235474-5G 235474-25G
220046-448EA 91.85 USD
220046-448 220046-447
Acridine orange base ≥75% (dye basis), Sigma-Aldrich®
Acridine orange
Formula: C₁₇H₁₉N₃
Storage Temperature: Ambient
MDL Number: MFCD00005029
CAS Number: 494-38-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=888823beccf53209a405a0fa2635aa17c7031fa8&vid=4d03cc4a6dbc866c2c25327340e9b2728c334a9a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR