JC-1 iodide salt ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, JC-1, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, (E)-5,6-Dichloro-2-(3-(5,6-dichloro-1,3-diethyl-1,3-dihydro-2H-benzo[d]imidazol-2-ylidene)prop-1-en-1-yl)-1,3-diethyl-1H-benzo[d]imidazol-3-ium iodide, CBIC2(3), 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide

22200 22201
76483-558EA 124.31 USD
76483-558 76483-560
JC-1 iodide salt ≥95%
JC-1 iodide salt

JC-1 is widely used for determining mitochondrial membrane potential with flow cytometry.

Formula: C₂₅H₂₇Cl₄IN₄
MW: 652.23 g/mol
MDL Number: MFCD00188475
CAS Number: 47729-63-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85d6d83390a09881c74f87212e58f13d0a014022&vid=3259ffd518b4bdf2fa9051a7c6c4d19e0d31e6e0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR