Calcein AM ≥95%, Ultra Pure Grade
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester
, Calcein acetoxymethyl ester
22004
22003
76483-468EA
240.68
USD
76483-468
76483-466
Calcein AM ≥95%, Ultra Pure Grade
Calcein AM
Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein.
Formula:
C₄₆H₄₆N₂O₂₃ MW: 994.86 g/mol |
MDL Number:
MFCD05861516 CAS Number: 148504-34-1 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...