Fura Red, AM ≥90%

Supplier: AAT BIOQUEST INC
Warning

21046 21048
76483-182EA 437.45 USD
76483-182 76483-184
Fura Red, AM ≥90%
Fura Red, AM

Fura Red is a visible light-excitable fura-2 analog that offers unique possibilities for ratiometric measurement of calcium ion in single cells by microphotometry, imaging or flow cytometry when used with single excitation, green-fluorescent calcium indicators.

Formula: C₄₇H₅₂N₄O₂₄S
CAS Number: 149732-62-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46aee770d2c30e7adb87a74cdb81d3ca4e72af55&vid=89ea585f356f82c089dd058d6923ced60b83cdc4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR