Indo-1 AM ≥95%, Ultra Pure Grade
Supplier: AAT Bioquest
|
Warning
|
21032
21036
76483-170EA
188.87
USD
76483-170
76483-172
Indo-1 AM ≥95%, Ultra Pure Grade
Indo-1 AM
Calcium measurements are critical for numerous biological investigations.
Formula:
C₄₇H₅₁N₃O₂₂ |
MDL Number:
MFCD00036955 CAS Number: 112926-02-0 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...