Fura-FF, pentapotassium salt [Fura-2FF, pentapotassium salt] ≥95%

Supplier: AAT BIOQUEST INC
Warning

21028 21029
76483-162EA 249.58 USD
76483-162 76483-164
Fura-FF, pentapotassium salt [Fura-2FF, pentapotassium salt] ≥95%
Fura-FF, pentapotassium salt [Fura-2FF, pentapotassium salt]

Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.

Formula: C₂₈H₁₈F₂K₅N₃O₁₄
MW: 853.94 g/mol
CAS Number: 192140-58-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a965dd5eef8f14c9c23051e872ca81f897573d49&vid=78ecc77af02fcd91ac75497dad14ea371f41aaf1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR