Fura 2-AM ≥95%, Ultra Pure Grade

Supplier: AAT BIOQUEST INC
Warning

Synonyms: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester

21021 21023
76483-152EA 188.87 USD
76483-152 76483-154
Fura 2-AM ≥95%, Ultra Pure Grade
Fura 2-AM

Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.

Formula: C₄₄H₄₇N₃O₂₄
MW: 1001.857 g/mol
MDL Number: MFCD00036976
CAS Number: 108964-32-5
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR