LDS 751 ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
LDS-751, 6-(Dimethylamino)-2-(4-(4-(dimethylamino)phenyl)buta-1,3-dien-1-yl)-1-ethylquinolin-1-ium perchlorate
17561
17567
76482-904EA
462.69
USD
76482-904
76482-906
LDS 751 ≥95%
LDS 751
LDS 751 is a fluorescent cell-permeant nucleic acid stain that can be well excited with 488 nm laser line although it has a peak excitation at ~543 nm on dsDNA.
Formula:
C₂₅H₃₀N₃·ClO₄ MW: 471.98 g/mol |
MDL Number:
MFCD00467069 CAS Number: 181885-68-7 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...