bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Bisbenzimide H 33258, 4-(5-(4-Methylpiperazin-1-yl)-1H,1'H-[2,5'-bibenzo[d]imidazol]-2'-yl)phenol trihydrochloride, 2'-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bibenzimidazole trihydrochloride hydrate

17520 17523
76482-868EA 124.31 USD
76482-868 76482-870
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥95%
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)

The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.

Formula: C₂₅H₂₄N₆O·3HCl·xH₂O
MW: 533.88 g/mol (anhy)
MDL Number: MFCD00012679
CAS Number: 23491-45-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b6851d2f3cca5b4f1783f3c851b9c9fdc1727de&vid=246c838bf7a86632b2ee16fe8bfde97a747a69ec&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR