4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride, DAPI 2HCl
17510
17511
17513
76482-850EA
96.26
USD
76482-850
76482-852
76482-854
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥95%
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)
DAPI is a fluorescent stain that binds strongly to DNA.
Formula:
C₁₆H₁₅N₅·2HCl MW: 350.25 g/mol |
MDL Number:
MFCD00012681 CAS Number: 28718-90-3 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...