Aminoallyl-dUTP sodium salt (aa-dUTP) 4 mM in TRIS buffer ≥95% (pH 7.5)

Supplier: AAT BIOQUEST INC
Warning

Synonyms: AA-dUTP, 5-[3-Aminoallyl]-2′-deoxyuridine 5′-triphosphate

17004 17005
76482-794EA 124.31 USD
76482-794 76482-796
Aminoallyl-dUTP sodium salt (aa-dUTP) 4 mM in TRIS buffer ≥95% (pH 7.5)
Aminoallyl-dUTP sodium salt (aa-dUTP)

The amine-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce amine-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.

Formula: C₁₂H₂₀N₃O₁₄P₃·xNa+
MW: 523.22 g/mol (free acid)
CAS Number: 936327-10-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=725c9fffa112cc3bc13da7b45361898fc514fbef&vid=a47e51d3224b1e3e4bf2c38b7a3650aa99eca628&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR