Dihydrorhodamine 123 ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate, 2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester

15206 15207
76482-328EA 124.31 USD
76482-328 76482-330
Dihydrorhodamine 123 ≥95%
Dihydrorhodamine 123

DihydrorH₂O₂damine 123 is by far the most-used probe for measurement of intracellular H₂O₂

Formula: C₂₁H₁₈N₂O₃
MDL Number: MFCD04221428
CAS Number: 109244-58-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=719b5faab3352ed6b9099af98a2f56186c421084&vid=abd6d6353b743c0a32e49a56870a23d5c7241211&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR