Resorufin alpha-D-glucopyranoside ≥95%

Supplier: AAT BIOQUEST INC
Warning

14022 14023
76482-304EA 476.9 USD
76482-304 76482-306
Resorufin alpha-D-glucopyranoside ≥95%
Resorufin alpha-D-glucopyranoside

Resorufin alpha-D-glucopyranoside is a sensitive fluorogenic substrate that generates a red fluorescent product (resorufin) upon interaction with alpha-glucosidase.

Formula: C₁₈H₁₇NO₈
CAS Number: 136565-96-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fd9292cd5b6d7901172a19aad971fbded3a471f&vid=3e53a4a8ee802236df3262415d7af8f8403865a1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR