4-Methylumbelliferyl-β-D-galactopyranoside ≥95%

Supplier: AAT BIOQUEST INC
Warning

14010 14012
76482-292EA 96.26 USD
76482-292 76482-294
4-Methylumbelliferyl-β-D-galactopyranoside ≥95%
4-Methylumbelliferyl-β-D-galactopyranoside

4-Methylumbelliferyl-beta-D-galactopyranoside (MUG) is a fluorogenic MU substrate for detecting galactosidase in cell extracts and purified enzyme preparations using a fluorescence microplate reader or fluorometer.

Formula: C₁₆H₁₈O₈
MW: 338.312 g/mol
MDL Number: MFCD00036773
CAS Number: 6160-78-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6982cdd7aea2a1c52eb54d8a45bfdb0f241e20c2&vid=e0c379a31bb4e03aa90e6fbee81396a6e178a523&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR