D-(-)-Luciferin sodium salt monohydrate ≥95%

Supplier: AAT BIOQUEST INC
Warning

12509 12510 12511
76482-000EA 96.26 USD
76482-000 76482-002 76482-004
D-(-)-Luciferin sodium salt monohydrate ≥95%
D(-)-Luciferin sodium salt

Luciferin is the most popular and versatile bioluminescent substrate.

Formula: C₁₁H₇N₂NaO₃S₂·H₂O
MW: 320.32 g/mol
MDL Number: MFCD11865368
CAS Number: 103404-75-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9545cea070c2a9fa67002828fa7e826c6e81e881&vid=4acb507e8831cd7b26a5d8d8d9b4ebd3ac5d9a55&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR