4-Methylumbelliferyl phosphate ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
11614
11617
76481-958EA
96.26
USD
76481-958
76481-960
4-Methylumbelliferyl phosphate ≥95%
4-Methylumbelliferyl phosphate
A fluorogenic substrate for phosphatases.
Formula:
C₁₀H₉O₆P |
MDL Number:
MFCD00016969 CAS Number: 3368-04-5 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...