6-FAM phosphoramidite [5'-Fluorescein phosphoramidite] ≥90%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
6-Fluorescein Phosphoramidite, 6-((6-(((2-Cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)hexyl)carbamoyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl bis(2,2-dimethylpropanoate), 5'-Fluorescein CE-Phosphoramidite (6-FAM)
6016
6017
76481-824EA
148
USD
76481-824
76481-826
6-FAM phosphoramidite [5'-Fluorescein phosphoramidite] ≥90%
6-FAM phosphoramidite [5'-Fluorescein phosphoramidite]
There are several ways of labeling an oligonucleotide with fluorescein.
Formula:
C₄₆H₅₈N₃O₁₀P MW: 843.94 g/mol |
MDL Number:
MFCD01940925 CAS Number: 204697-37-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...