Fmoc-Lys(Dabcyl)-OH ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Nepsilon-4-[4-(Dimethylamino)phenylazo]benzoyl-Nα-Fmoc-L-lysine

5040 5041
76481-730EA 124.31 USD
76481-730 76481-732
Fmoc-Lys(Dabcyl)-OH ≥90%
Fmoc-Lys(Dabcyl)-OH

FMOC-Lys(DABCYL)-OH is a building block for in-sequence Lys labeling by DABCYL.

Formula: C₃₆H₃₇N₅O₅
MW: 619.722 g/mol
MDL Number: MFCD00467694
CAS Number: 146998-27-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1477885921ad694d8dabba053c13991781862b6c&vid=b0fb7ea94e726f9d83f44f136859b1b812ad5de3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR