Fmoc-Asp(Edans)-OH ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Fmoc-Asp(EDANS)-OH

5000 5001 5002
76481-700EA 124.31 USD
76481-700 76481-702 76481-704
Fmoc-Asp(Edans)-OH ≥90%
Fmoc-Asp(Edans)-OH

FMOC-Asp(EDANS)-OH is a building block for in-sequence peptide Asp labeling by EDANS.

Formula: C₃₁H₂₉N₃O₈S
MW: 603.646 g/mol
MDL Number: MFCD02094140
CAS Number: 182253-73-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac5f20d3a345335247639abbfb5ce08925564831&vid=523a569de809d7174d42ff94a178b5a38b3cdce3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR