Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulphonate, Sulphosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulpho-SMCC Sodium Salt, SMCC, Water-Soluble, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, sulpho-SMCC, Sulfo-SMCC

4504 4505 4506
76481-690EA 311.76 USD
76481-690 76481-692 76481-694
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥90%
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

Sulfo-SMCC is the water-soluble version of SMCC.

Formula: C₁₆H₁₈N₂O₉S
MDL Number: MFCD00054978
CAS Number: 92921-24-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fe24ecb8d003df1ec7fe314370bca9891576fa9&vid=87f000e8cab7c3fa2be70b7c58ca2a7ac0d92dde&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR