N-Boc-ethylenediamine
Supplier: AAT BIOQUEST INC
|
Warning
|
4013
4014
76481-666EA
500.94
USD
76481-666
76481-668
N-Boc-ethylenediamine
N-Boc-ethylenediamine
N-BOC-ethylenediamine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry.
Formula:
C₇H₁₆N₂O₂ |
MDL Number:
MFCD00191871 CAS Number: 57260-73-8 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...