N-(tert-Butoxycarbonyl)-1,5-diaminopentane

Supplier: AAT BIOQUEST INC
Warning

Synonyms: N-Boc-1,5-diaminopentane, tert-Butyl N-(5-Aminopentyl)carbamate, tert-Butyl (5-aminopentyl)carbamate, N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, NH2-C5-NH-Boc, N-Boc-1,5-pentanediamine

4010 4011
76481-662EA 947.71 USD
76481-662 76481-664
N-(tert-Butoxycarbonyl)-1,5-diaminopentane
N-(tert-Butoxycarbonyl)-1,5-diaminopentane

N-BOC-cadaverine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry.

Formula: C₁₀H₂₂N₂O₂
MW: 202.30 g/mol
MDL Number: MFCD00210020
CAS Number: 51644-96-3
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ce7e2b09f4b90a4135607939b5fa0d69c6985d1&vid=a5a7f2b7df191c9ccf61b4754fad5069439528d6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR