N-FMOC-ethylenediamine ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
4006
4007
76481-658EA
626.51
USD
76481-658
76481-660
N-FMOC-ethylenediamine ≥95%
N-FMOC-ethylenediamine
N-FMOC-ethylenediamine is a monoprotected diamine that is widely used as a building block in bioconjugate chemistry.
Formula:
C₁₇H₁₈N₂O₂ MW: 282.34 g/mol |
CAS Number:
166410-32-8 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...